ADVH49B82812 AChemBlock

methyl 2-{bis[(tert-butoxy)carbonyl]amino}pyrimidine-5-carboxylate

  • Molecular Weight : 353.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-bis-tert-butoxy-carbonyl-amino-pyrimidine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84942
Availability details for
Quantity
  • Availability
  • Add to Cart