ADVH49B82814 AChemBlock

methyl 2-(1-methylazetidin-2-yl)acetate;hydrochloride

  • Molecular Weight : 179.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-1-methylazetidin-2-yl-acetate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84955
Availability details for
Quantity
  • Availability
  • Add to Cart