ADVH49B82815 AChemBlock

methyl 1H,2H,3H-pyrrolo[3,2-c]pyridine-7-carboxylate

  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1h-2h-3h-pyrrolo-3-2-c-pyridine-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84957
Availability details for
Quantity
  • Availability
  • Add to Cart