ADVH49B82817 AChemBlock

methyl 1,5,5-trimethyl-4-oxo-1H,4H,5H,6H-cyclopenta[b]pyrrole-2-carboxylate

  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-5-5-trimethyl-4-oxo-1h-4h-5h-6h-cyclopenta-b-pyrrole-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84961
Availability details for
Quantity
  • Availability
  • Add to Cart