ADVH49B82821 AChemBlock

ethyl trans-2-amino-5,5-difluoro-cyclohexanecarboxylate

  • Molecular Weight : 207.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-trans-2-amino-5-5-difluoro-cyclohexanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85015
Availability details for
Quantity
  • Availability
  • Add to Cart