ADVH49B82824 AChemBlock

ethyl cis-4-(trifluoromethyl)pyrrolidine-3-carboxylate hydrochloride

  • Molecular Weight : 247.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-cis-4-trifluoromethyl-pyrrolidine-3-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85018
Availability details for
Quantity
  • Availability
  • Add to Cart