ADVH49B82828 AChemBlock

ethyl 5-amino-3-benzyloxy-1-methyl-2-oxo-pyridine-4-carboxylate

  • Molecular Weight : 302.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-amino-3-benzyloxy-1-methyl-2-oxo-pyridine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85026
Availability details for
Quantity
  • Availability
  • Add to Cart