ADVH49B82829 AChemBlock

ethyl 4-(cyclopropylamino)-2-[4-(4-ethylsulfonylpiperazin-1-yl)anilino]pyrimidine-5-carboxylate

  • Molecular Weight : 474.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-cyclopropylamino-2-4-4-ethylsulfonylpiperazin-1-yl-anilino-pyrimidine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85030
Availability details for
Quantity
  • Availability
  • Add to Cart