ADVH49B8282B AChemBlock

ethyl 3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carboxylate

  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-4-dimethoxybicyclo-4-2-0-octa-1-3-5-triene-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85035
Availability details for
Quantity
  • Availability
  • Add to Cart