ADVH49B82830 AChemBlock

ethyl 2-(3-amino-1-benzhydryl-azetidin-3-yl)-2,2-difluoro-acetate

  • Molecular Weight : 360.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-3-amino-1-benzhydryl-azetidin-3-yl-2-2-difluoro-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85044
Availability details for
Quantity
  • Availability
  • Add to Cart