ADVH49B82832 AChemBlock

ethyl (3S,4R)-3-(2-hydroxyethyl)-4-(hydroxymethyl)pyrrolidine-1-carboxylate

  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3s-4r-3-2-hydroxyethyl-4-hydroxymethyl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85050
Availability details for
Quantity
  • Availability
  • Add to Cart