ADVH49B82835 AChemBlock

ethyl (1S,2S,4R)-rel-4-amino-2-methyl-cyclohexanecarboxylate

  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1s-2s-4r-rel-4-amino-2-methyl-cyclohexanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85058
Availability details for
Quantity
  • Availability
  • Add to Cart