ADVH49B82837 AChemBlock

ethyl (1R,3S,4R)-4-(tert-butoxycarbonylamino)-3-fluoro-cyclohexanecarboxylate

  • Molecular Weight : 289.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1r-3s-4r-4-tert-butoxycarbonylamino-3-fluoro-cyclohexanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85062
Availability details for
Quantity
  • Availability
  • Add to Cart