ADVH49B82839 AChemBlock

cis-4-fluoro-3-methyl-piperidine

  • Molecular Weight : 117.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-4-fluoro-3-methyl-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85082
Availability details for
Quantity
  • Availability
  • Add to Cart