ADVH49B8283A AChemBlock

cis-4-(1-hydroxy-1-methyl-ethyl)cyclohexanecarboxylic acid

  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-4-1-hydroxy-1-methyl-ethyl-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85084
Availability details for
Quantity
  • Availability
  • Add to Cart