ADVH49B8283B AChemBlock

cis-1-ethynyl-4-fluoro-cyclohexane

  • Molecular Weight : 126.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-1-ethynyl-4-fluoro-cyclohexane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85098
Availability details for
Quantity
  • Availability
  • Add to Cart