ADVH49B8283C AChemBlock

cis-1-(bromomethyl)-4-fluorocyclohexane

  • Molecular Weight : 195.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-1-bromomethyl-4-fluorocyclohexane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85102
Availability details for
Quantity
  • Availability
  • Add to Cart