ADVH49B8283E AChemBlock

benzyl trans-1-amino-3-methoxycyclobutanecarboxylate hydrochloride

  • Molecular Weight : 271.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-trans-1-amino-3-methoxycyclobutanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85106
Availability details for
Quantity
  • Availability
  • Add to Cart