ADVH49B82841 AChemBlock

benzyl exo-(1S,5R)-3-(methylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate

  • Molecular Weight : 274.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-exo-1s-5r-3-methylamino-8-azabicyclo-3-2-1-octane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85123
Availability details for
Quantity
  • Availability
  • Add to Cart