ADVH49B82842 AChemBlock

benzyl cis-1-amino-3-ethoxycyclobutanecarboxylate hydrochloride

  • Molecular Weight : 285.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-cis-1-amino-3-ethoxycyclobutanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85128
Availability details for
Quantity
  • Availability
  • Add to Cart