ADVH49B82843 AChemBlock

benzyl 1-(benzyloxycarbonylamino)-3-oxo-cyclobutanecarboxylate

  • Molecular Weight : 353.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-1-benzyloxycarbonylamino-3-oxo-cyclobutanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85136
Availability details for
Quantity
  • Availability
  • Add to Cart