ADVH49B82844 AChemBlock

acetamido 1-ethyl-2-oxo-1,2,3,4-tetrahydroquinoline-3-carboxylate

  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/acetamido-1-ethyl-2-oxo-1-2-3-4-tetrahydroquinoline-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85155
Availability details for
Quantity
  • Availability
  • Add to Cart