ADVH49B82846 AChemBlock

8-tert-butoxycarbonyl-2-hydroxy-8-azaspiro[4.5]decane-4-carboxylic acid

  • Molecular Weight : 299.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-tert-butoxycarbonyl-2-hydroxy-8-azaspiro-4-5-decane-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85161
Availability details for
Quantity
  • Availability
  • Add to Cart