ADVH49B82851 AChemBlock

8,8-difluoro-5-oxa-2-azaspiro[3.5]nonane hydrochloride

  • Molecular Weight : 199.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-8-difluoro-5-oxa-2-azaspiro-3-5-nonane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85181
Availability details for
Quantity
  • Availability
  • Add to Cart