ADVH49B82852 AChemBlock

7-oxaspiro[3.5]nonane-9-carboxylic acid

  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-oxaspiro-3-5-nonane-9-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85185
Availability details for
Quantity
  • Availability
  • Add to Cart