ADVH49B82853 AChemBlock

7-oxaspiro[3.5]nonan-9-one

  • Molecular Weight : 140.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-oxaspiro-3-5-nonan-9-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85186
Availability details for
Quantity
  • Availability
  • Add to Cart