ADVH49B8285D AChemBlock

7-bromo-2H,3H,4H,5H-pyrido[4,3-f][1,4]oxazepin-5-one

  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2h-3h-4h-5h-pyrido-4-3-f-1-4-oxazepin-5-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85209
Availability details for
Quantity
  • Availability
  • Add to Cart