ADVH49B82862 AChemBlock

6-oxaspiro[3.4]octan-8-one

  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-oxaspiro-3-4-octan-8-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85223
Availability details for
Quantity
  • Availability
  • Add to Cart