ADVH49B82864 AChemBlock

6-oxa-2-azaspiro[3.4]octane-8-carboxylic acid

  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-oxa-2-azaspiro-3-4-octane-8-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85225
Availability details for
Quantity
  • Availability
  • Add to Cart