ADVH49B82869 AChemBlock

6-iodo-3-methyl-5-oxo-thiazolo[3,2-a]pyridine-8-carbonitrile

  • Molecular Weight : 316.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-iodo-3-methyl-5-oxo-thiazolo-3-2-a-pyridine-8-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85233
Availability details for
Quantity
  • Availability
  • Add to Cart