ADVH49B8286D AChemBlock

6-chloro-3H-imidazo[4,5-c]pyridine-2-carbaldehyde

  • Molecular Weight : 181.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-3h-imidazo-4-5-c-pyridine-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85249
Availability details for
Quantity
  • Availability
  • Add to Cart