ADVH49B82870 AChemBlock

6-Bromo-2-fluoro-3-(methoxymethoxy)phenylboronic acid pinacol ester

  • Molecular Weight : 379.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-fluoro-3-methoxymethoxy-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70480
Availability details for
Quantity
  • Availability
  • Add to Cart