ADVH49B82872 AChemBlock

6-azabicyclo[3.1.1]heptane hydrochloride

  • Molecular Weight : 133.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-azabicyclo-3-1-1-heptane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85261
Availability details for
Quantity
  • Availability
  • Add to Cart