ADVH49B82878 AChemBlock

5-tert-butoxycarbonyl-3-oxo-2,5-diazaspiro[3.4]octane-7-carboxylic acid

  • Molecular Weight : 270.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-tert-butoxycarbonyl-3-oxo-2-5-diazaspiro-3-4-octane-7-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85280
Availability details for
Quantity
  • Availability
  • Add to Cart