ADVH49B82880 AChemBlock

5-Fluoro-4-formyl-3-methoxyphenylboronic acid pinacol ester

  • Molecular Weight : 298.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-4-formyl-3-methoxyphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70580
Availability details for
Quantity
  • Availability
  • Add to Cart