ADVH49B82881 AChemBlock

5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

  • Molecular Weight : 262.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85300
Availability details for
Quantity
  • Availability
  • Add to Cart