ADVH49B82889 AChemBlock

5-bromonorpinan-1-amine

  • Molecular Weight : 190.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromonorpinan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85318
Availability details for
Quantity
  • Availability
  • Add to Cart