ADVH49B8288D AChemBlock

5-bromo-3-fluoro-imidazo[1,2-a]pyridine

  • Molecular Weight : 215.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-fluoro-imidazo-1-2-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85330
Availability details for
Quantity
  • Availability
  • Add to Cart