ADVH49B8288E AChemBlock

5-Bromo-3-chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester

  • Molecular Weight : 383.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-chloro-2-fluoro-4-methoxyphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70852
Availability details for
Quantity
  • Availability
  • Add to Cart