ADVH49B8288F AChemBlock

5-Bromo-3-(octyloxy)phenylboronic acid pinacol ester

  • Molecular Weight : 429.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-octyloxy-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70803
Availability details for
Quantity
  • Availability
  • Add to Cart