ADVH49B82890 AChemBlock

5-Bromo-2-chlorophenylboronic acid pinacol ester

  • Molecular Weight : 335.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-chlorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70623
Availability details for
Quantity
  • Availability
  • Add to Cart