ADVH49B82891 AChemBlock

5-bromo-2-[(E)-1-fluoro-2-iodo-vinyl]pyridine

  • Molecular Weight : 327.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-e-1-fluoro-2-iodo-vinyl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85332
Availability details for
Quantity
  • Availability
  • Add to Cart