ADVH49B82893 AChemBlock

5-bromo-2,2-difluoro-spiro[2.3]hexane

  • Molecular Weight : 197.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-2-difluoro-spiro-2-3-hexane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85333
Availability details for
Quantity
  • Availability
  • Add to Cart