ADVH49B82894 AChemBlock

5-bromo-2-(trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine

  • Molecular Weight : 265.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-trifluoromethyl-1h-pyrrolo-3-2-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85334
Availability details for
Quantity
  • Availability
  • Add to Cart