ADVH49B82895 AChemBlock

5-borononicotinic acid pinacol ester

  • Molecular Weight : 267.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-borononicotinic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85339
Availability details for
Quantity
  • Availability
  • Add to Cart