ADVH49B82898 AChemBlock

5,5-difluorotetrahydropyran-3-carboxylic acid

  • Molecular Weight : 166.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-5-difluorotetrahydropyran-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85350
Availability details for
Quantity
  • Availability
  • Add to Cart