ADVH49B82899 AChemBlock

5,5-difluoro-5H,6H,7H,8H-pyrido[3,4-d]pyrimidine hydrochloride

  • Molecular Weight : 207.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-5-difluoro-5h-6h-7h-8h-pyrido-3-4-d-pyrimidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85352
Availability details for
Quantity
  • Availability
  • Add to Cart