ADVH49B8289C AChemBlock

5-(Dimethylamino)-2-methylphenylboronic acid pinacol ester

  • Molecular Weight : 279.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-dimethylamino-2-methylphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70670
Availability details for
Quantity
  • Availability
  • Add to Cart