ADVH49B8289D AChemBlock

5-(bromomethyl)-2-(trifluoromethyl)tetrahydropyran

  • Molecular Weight : 247.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromomethyl-2-trifluoromethyl-tetrahydropyran.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85358
Availability details for
Quantity
  • Availability
  • Add to Cart