ADVH49B8289E AChemBlock

4-methyl-2-oxo-1,2,3,4-tetrahydroquinoxaline-6-boronic acid pinacol ester

  • Molecular Weight : 306.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-methyl-2-oxo-1-2-3-4-tetrahydroquinoxaline-6-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85368
Availability details for
Quantity
  • Availability
  • Add to Cart